Anti-POLR2A Mouse Monoclonal Antibody

Supplier: Rockland Immunochemical
200-301-V00
75934-112EA 586.12 USD
75934-112
Anti-POLR2A Mouse Monoclonal Antibody
Antibodies
RNA polymerase II (pol II) is a key enzyme in the regulation and control of gene transcription. It is able to unwind the DNA double helix, synthesize RNA, and proofread the result. Pol II is a complex enzyme, consisting of 12 subunits, of which the B1 subunit (UniProt/Swiss-Prot entry P24928) is the largest. Together with the second largest subunit, B1 forms the catalytic core of the RNA polymerase II transcription machinery. Anti-Pol II S2p Antibody is ideal for research in Gene Expression, Transcription, and Genetics.

Anti-Pol II pS2 Antibody is suitable for Chromatin Immunoprecipitation Sequencing, Chromatin Immunoprecipitation, ELISA, Immunofluorescence and Western Blots. Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 225 kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: POLR2A
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR