OXSR1
Supplier: Rockland Immunochemical
009-001-S58
75934-034EA 364 USD
75934-034 75934-036
OXSR1
Proteins and Peptides
OSR1 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
OSR1 is a member of the Ser/Thr family of protein kinases. OSR1 is activated by osmotic stresses and regulates the downstream protein kinase such as PAK1 in order to establish the sensor/signaling modules that initiate the cellular response to environmental stress (1). OSR1 can be phosphorylated by ATM or ATR during DNA damage process (2). OSR1 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cardiovascular Disease, and Ser/Thr Kinases research.
OSR1 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. OSR1 Protein is suitable for use in Western Blot. Expect a band approximately ~86kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.