Anti-NUP155 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-DF4
75934-024EA 635.22 USD
75934-024
Anti-NUP155 Rabbit Polyclonal Antibody
Antibodies
The nuclear pore complex (NPC) is a protein assembly localized at the nuclear rim and mediates macromolecular transport between the nucleus and the cytoplasm. One protein in this assembly is the nucleoporin (NUP)-155, which is localized symmetrically to both the nucleoplasmic and cytoplasmic faces of the NPC. NUP155 has been reported to interact with both NUP35 and the mRNA export factor Gle1, but the functions of these interactions are still unknown. NUP155 has also been shown to be required for NPC assembly and nuclear envelope (NE) membrane fusion. NUP155 is recruited late in NE formation, suggesting that NUP155 defines an essential late step in NE assembly. NUP155 has recently been identified as an HIV dependency factor (HDF), suggesting that NUP155 may be an important drug target in HIV treatment. At least two isoforms of NUP155 are known to exist.

Anti-NUP155 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 155 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NUP155
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR