Human Recombinant LIF
Supplier: Rockland Immunochemical
009-001-V72-0025 009-001-V72-0005 009-001-V72-0100
75933-808EA 352.09 USD
75933-808 75933-810 75934-846 75934-850
Human Recombinant LIF
Proteins and Peptides
Recombinant Human LIF control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Human Leukemia Inhibitory Factor (LIF) is member of the IL-6 family and is made by a variety of tissues. Human LIF signals through gp130 to activiate STAT3 in human ESCs . In mouse ESCs, LIF promotes self-renewal and pluripotentcy, but in human cells there is no indication that LIF has the same function. Instead, FGF-basic (FGF-2) and Activin A are used to promote self-renewal and pluripotent potential. Recombinant human LIF is a non-glycosylated protein, containing 181 amino acids, with a molecular weight of 19.9 kDa.
Leukemia Inhibitory Factor Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Leukemia Inhibitory Factor in immunological assays.