Anti-WNVgp1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-FY4
75933-618EA 635.22 USD
75933-618
Anti-WNVgp1 Rabbit Polyclonal Antibody
Antibodies
West Nile Virus (WNV) is a member of the Flaviviridae, a plus-stranded virus family that includes St. Louis encephalitis virus, yellow fever virus, and Dengue virus. WNV was initially isolated in 1937 in the West Nile region of Uganda and has become prevalent in Africa, Asia, and Europe. It has rapidly spread across the United States with cases being observed in every continental state. Virus particles consist of a dense core made up of the core/capsid protein encapsulating the RNA genome surrounded by a membrane envelope embedded with envelope and matrix proteins. While the viral core protein is thought to contribute to the WNV-associated inflammation via apoptosis induced though the caspase-9 pathway, the highly glycosylated envelope protein plays a major role for WNV entry into target cells as this entry can be inhibited by using a recombinant domain III from the envelope glycoprotein. The WNV receptor has recently been identified as alpha v beta 3 integrin.

Anti-West Nile Virus Envelope Antibody has been tested for use in ELISA. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 380 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: WNVgp1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR