Anti-VKORC1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-FW9
75933-582EA 635.22 USD
75933-582
Anti-VKORC1 Rabbit Polyclonal Antibody
Antibodies
Vitamin K epoxide reductase complex subunit 1 (VKORC1) is the enzyme that is responsible for reducing vitamin K 2,3-epoxide to the enzymatically activated form which is essential for blood clotting. This enzymatically activated form of vitamin K is a reduced form required for the carboxylation of glutamic acid residues in some blood-clotting proteins. Fatal bleeding can be caused by vitamin K deficiency and by the vitamin K antagonist warfarin, and it is VKORC1 that is sensitive to warfarin. In humans, mutations in this gene can be associated with deficiencies in vitamin-K-dependent clotting factors and, in humans and rats, with warfarin resistance.

Anti-VKORC1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 18 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: VKORC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR