UBE2B
Supplier: Rockland Immunochemical
009-001-U28S
75933-308EA 228.81 USD
75933-308 75933-310
UBE2B
Proteins and Peptides
UBE2B control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
UBE2B or ubiquitin-conjugating enzyme E2B is a member of the E2 ubiquitin-conjugating enzyme family which is required for post-replicative DNA damage repair. UBE2B is specifically important during chronic low-dose ultraviolet exposure to prevent counterproductive DNA checkpoint activation and allow cells to proliferate normally (1). The activity of UBE2B is required within the synaptonemal complex and for meiotic recombination in spermatocytes (2). Experimental inactivation of the UBE2B gene in mice causes male infertility. UBE2B Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Cancer, Cell Cycle, and Neurobiology research.
UBE2B Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. UBE2B Protein is suitable for use in Western Blot. Expect a band approximately ~17kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.