Anti-TTYH2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-FS7
75933-204EA 635.22 USD
75933-204
Anti-TTYH2 Rabbit Polyclonal Antibody
Antibodies
TTYH2 is a member of the tweety family of proteins, a family of chloride anion channels containing five transmembrane regions. TTYH2 is a Ca2+-activated large conductance chloride (Cl-) channel and may play a role in kidney tumorigenesis. It is also upregulated in colon carcinoma and is thought to play a role in cell proliferation and call aggregation, suggesting that TTYH2 may play a role in regulating both proliferation and metastatic potentials of colorectal cancer. The ubiquitin-protein ligase Nedd4-2 binds to and ubiquitinates TTYH2, thereby regulating its cell surface expression and total cellular levels.

Anti-TTYH2 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 59 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TTYH2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR