Anti-TTBK1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-FS3
75933-196EA 635.22 USD
75933-196
Anti-TTBK1 Rabbit Polyclonal Antibody
Antibodies
Tau tubulin kinase (TTBK1) belongs to the casein kinase 1 superfamily and is involved in the phosphorylation of specific serine/threonine residues in paired helical filaments of the tau protein. It is specifically expressed in the brain and induces tau aggregation in human neuronal cells in a dose-dependent manner. TTBK1 levels have been found to be upregulated in the brains of Alzheimer's disease (AD) patients, and mice expressing human TTBK1 protein showed significant age-dependent memory impairment. These mice displayed increased levels of the CDK5 activators p25 and p35, and reduced levels of the NMDA receptor types 2B and 2D, suggesting that TTBK1 may play a role in memory dysfunction in AD patients. Multiple isoforms of TTBK1 are known to exist. This antibody is predicted to not cross-react with TTBK2.

Anti-TTBK1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 143 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TTBK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR