Anti-TNNI3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-X32
75933-146EA 656.12 USD
75933-146
Anti-TNNI3 Rabbit Polyclonal Antibody
Antibodies
Troponin I (cTnI) is 1 of 3 subunits, along with troponin C (TnC) and troponin T (TnT) of troponin complex found in cardiac muscle. cTnI binds to actin in thin myofilaments to hold the troponin-tropomyosin complex in place, and when cTnI is phosphorylated by protein kinase C and protein kinase A at Ser23/24 it causes regulation of Ca2+- stimulated ATPase. Evidence suggests that phosphorylation of both serines 23 and 24 is required for the reduction in Ca2+-sensitivity and beneficial for relaxation of the heart. Ser23/24 phosphorylation is important for enhanced relaxation in response to prolonged activation of protein kinase C by endothelin in intact myocytes, while Thr144 plays an important role in the acute acceleration of relaxation. Troponin I pS23/24 Antibody is ideal for researchers interested in cancer research.

Anti-Phospho-Ser23/24 Troponin (cardiac) Antibody is stored with 100 μg per ml BSA. It is suitable for Western Blots and is specific for the ~25 kDa cardiac troponin I protein phosphorylated at Ser 23/24. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: TNNI3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR