Anti-Transferrin Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
209-4134-0100
75933-046EA 551.64 USD
75933-046
Anti-Transferrin Rabbit Polyclonal Antibody
Antibodies
Transferrin Antibody detects transferrin protein. Transferrins are iron binding transport proteins which can bind two Fe3+ ions in association with the binding of an anion, usually bicarbonate. It is responsible for the transport of iron from sites of absorption and heme degradation to those of storage and utilization. Serum transferrin may also have a further role in stimulating cell proliferation. Anti-transferrin Antibody is ideal for investigators involved in Cell Signaling, Neuroscience and Signal Transduction research.

Anti-Transferrin Antibody is suitable for western blotting, ELISA and IHC. Researchers should determine optimal titers for applications that are not stated below.

Type: Primary
Antigen: Transferrin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR