Anti-TLR6 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-FG3
75932-930EA 635.22 USD
75932-930
Anti-TLR6 Rabbit Polyclonal Antibody
Antibodies
Toll-like receptors (TLRs) are evolutionarily conserved pattern-recognition molecules resembling the toll proteins that mediate antimicrobial responses in Drosophila. These proteins recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses. The TLRs act through adaptor molecules such as MyD88 and TIRAP to activate various kinases and transcription factors so the organism can respond to potential infection. TLR6 was first identified as a close homolog of TLR1, sharing 69% sequence identify. Like TLR1, TLR6 can form heterodimers with TLR2, and these TLR6:TLR2 dimers coordinate macrophage activation by Gram-positive bacteria and the yeast cell wall particle zymosan. Activation of these complexes not only initiates pro-inflammatory cascades, but also can lead to apoptotic responses.

Anti-TLR6 Antibody has been tested for use in ELISA, Western Blotting, and Immunohistochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 92 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TLR6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR