Anti-TFEB Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-FD1
75932-844EA 635.22 USD
75932-844
Anti-TFEB Rabbit Polyclonal Antibody
Antibodies
The Transcription factor EB (TFEB) is a member in the basic helix-loop-helix leucine zipper superfamily of transcription factors that is translocated in a subset of renal tumors (1,2). Recent studies have shown that lysosomal biogenesis is regulated by TFEB (3), which is in turn regulated by the mammalian target of rapamycin (mTOR) complex 1 (4). Other evidence suggests that TFEB coordinates the major steps of the autophagic pathway by driving the expression of autophagy and lysosomal genes (5).

Anti-TFEB Antibody has been tested for use in ELISA, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 53 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TFEB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR