MAPT
Supplier: Rockland Immunochemical
009-001-T72S
75932-620EA 364.63 USD
75932-620 75932-622
MAPT
Proteins and Peptides
Tau-316 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Tau-316 encodes the microtubule-associated protein tau (MAPT) whose transcript undergoes complex, regulated alternative splicing, giving rise to several mRNA species. Tau-316 transcripts are differentially expressed in the nervous system, depending on stage of neuronal maturation and neuron type (1). Tau-316 gene mutations have been associated with several neurodegenerative disorders such as Alzheimer's disease, Pick's disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy (2). Tau-316 Protein is ideal for investigators involved in Signaling Proteins, Tau Proteins, Invasion/Metastasis, Neurobiology, and p38 Pathway research.
Tau-316 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. Tau-316 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~39kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.