Swine Serum Sterile, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D117-00-0500 D117-00-1000 D117-00-0100 D117-00-0050
75932-512EA 1043.73 USD
75932-512 75932-522 75932-524 75932-526
Swine Serum Sterile, Rockland Immunochemicals
Sera (Serum)
Sterile Serum
  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Swine Serum is used as a supplement to cell culture media. Swine Serum is also suitable for use as a component of bioassays, immunoassays or enzyme assays. Swine Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation. Swine Serum is ideal for investigators in Cancer, Immunology, and Cell Biology research.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal