Anti-NS1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-EX7
75932-408EA 635.22 USD
75932-408
Anti-NS1 Rabbit Polyclonal Antibody
Antibodies
Influenza A virus is a major public health threat, killing more than 30, 000 people per year in the USA. In early 2009, a novel swine-origin influenza A (H1N1) virus (S-OIV) was identified in specimens obtained from patients in Mexico and the United States. One of the less studied proteins encoded by, but not incorporated in, the influenza virus is the nonstructural protein (NS) 1. NS1 counters cellular antiviral activities and acts as a virulence factor. It can bind to double-stranded RNA and sequester it from 2'-5'OAS, preventing the activation of the RNAse L, which normally acts to degrade RNA and prevent virus replication. NS1 also binds to and inhibits the anti-viral protein kinase PKR.

Anti-Swine H1N1 Nonstructural Protein 1 Antibody has been tested for use in ELISA. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 25 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR