Swine Blood In Sodium Citrate, Rockland Immunochemicals
Supplier: Rockland Immunochemical
R217-0050
75932-390EA 388.66 USD
75932-390
Swine Blood In Sodium Citrate, Rockland Immunochemicals
Blood
Swine Blood In Sodium Citrate
  • aseptically collected and prepared to order
  • blood draw in Anticoagulant
Citrated swine blood is aseptically collected and prepared to order from our colony of animals. These cell suspensions are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Swine red blood cells are perishable and are collected and processed upon receipt of your order.
Washed pooled cells preparations, complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents.