Swine Blood Defibrinated, Rockland Immunochemicals
Supplier: Rockland Immunochemical
R117-0050
75932-386EA 372 USD
75932-386
Swine Blood Defibrinated, Rockland Immunochemicals
Blood
Swine Blood Defibrinated
  • aseptically collected and prepared to order
  • Blood agar plate preparations
This item is perishable and is collected and processed upon receipt of your order.