Anti-SUZ12 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-EX2
75932-380EA 635.22 USD
75932-380
Anti-SUZ12 Rabbit Polyclonal Antibody
Antibodies
The Polycomb group (PcG) genes contribute to the maintenance of cell identity, cell cycle regulation and oncogenesis (1). SUZ12 (suppressor of zeste 12) is a 90 - 95 kDa member of the polycomb group of transcriptional regulators (2). It is part of a transcription repression complex termed PRC2 (Polycomb repressive complex 2) that methylates histones in the nucleus, resulting in homeotic (pattern-inducing) gene silencing. It contains one C2H2-type zinc finger region and a C-terminal VEFS-box (3,4). SUZ12 is overexpressed in breast and colon cancer. SUZ12 may be a cause of endometrial stromal tumors (5).

Anti-SUZ12 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 83 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SUZ12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR