Mouse SDF-1 Beta/CXCL12
Supplier: Rockland Immunochemical
010-001-W23-0010 010-001-W23-0100
75932-346EA 4582 USD
75932-346 75932-350 75932-354 75932-358
Mouse SDF-1 Beta/CXCL12
Proteins and Peptides
Recombinant Mouse SDF-1 Beta/CXCL12 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Stromal Derived Growth Factor-1β (SDF-1β), also called CXCL12, is one of two splice variants made by a wide variety of cells when stimulated by inflammatory cytokines such as, TNF, IL-1 or LPS. SDF-1β signals through the G protein-couple receptor, CXCR4, to recruit activated leukocytes. Human SDF-1β shares 100% identity to feline SDF-1β. Recombinant mouse SDF-1β is a non-glycosylated protein, containing 72 amino acids, with a molecular weight of 8.5 kDa.
Stromal Cell-Derived Factor-1 beta Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Stromal Cell-Derived Factor-1 beta in immunological assays.