STAT2
Supplier: Rockland Immunochemical
009-001-T52S
75932-184EA 409.55 USD
75932-184 75932-186
STAT2
Proteins and Peptides
STAT2 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
STAT2 (Signal transducer and activator of transcription) is a family member of STAT transcription factors that are implicated in programming gene expression in biological events as diverse as embryonic development, programmed cell death, organogenesis, innate immunity, adaptive immunity and cell growth regulation (1). JAKs can phosphorylate and activate STATs. Activation of the JAK/STAT pathway in response to cytokine action can lead to regulation of gene expression during proliferation and differentiation of the immune cells. STATs have been shown to be involved in inflammation, autoimmune and allergic diseases (2). STAT2 Protein is ideal for investigators involved in Signaling Proteins, Transcription Proteins, Apoptosis/Autophagy, Cancer, ERK/MAPK Pathway, Inflammation, and JAK/STAT Pathway research.
STAT2 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. STAT2 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~138kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.