Anti-SPATA6 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-ET1
75932-140EA 635.22 USD
75932-140
Anti-SPATA6 Rabbit Polyclonal Antibody
Antibodies
The development of spermatogonial stem cells into spermatozoa, i.e., spermatogenesis, is a highly ordered process that requires multiple proteins, including some that are testis-specific or testis-specific isoforms of genes that are expressed in other tissues. One such protein is SPATA6: while its full-length isoform is expressed in multiple tissues including thymus, brain and testis, the shorter isoform is expressed only in testis. It has some homology to the motor domain of kinesin related proteins and with the C. elegans neural calcium sensor protein NCS-2.

Anti-SPATA6 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 56 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SPATA6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR