Anti-SPATA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-ES6
75932-130EA 635.22 USD
75932-130
Anti-SPATA1 Rabbit Polyclonal Antibody
Antibodies
SPATA1, a novel cell-surface protein, is involved in shaping the sperm head during spermatogenesis. SPATA1 is localized at chromosome 1q21. At least three isoforms of SPATA1 are known to exist. The longest is preferentially expressed in hematopoietic tissues. Its extracellular domain contains a single immunoglobulin-like domain, and its intracellular segment has two immunoreceptor tyrosine-based inhibition motifs (ITIMs). It is thought to have an important role in hematopoietic cell signaling. The mid-sized isoform contains a short intracellular part without ITIMs, while the shortest isoform lacks the transmembrane segment and represents a potential soluble protein (2, 3).

Anti-SPATA1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 50 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SPATA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR