Anti-SOX4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-V96
75932-124EA 498 USD
75932-124
Anti-SOX4 Rabbit Polyclonal Antibody
Antibodies
SOX4 belongs to the SOX family of transcription factors. These transcription factors are characterized by the conserved HMG (high mobility group) DNA binding domain and are involved in embryonic development. SOX4 has been shown to be important for the development of the cardiac outflow tract and of the internal nervous system. SOX 4 may also be involved in the apoptosis pathway were it can either lead to cell death or to tumorigenesis, and has been associated with several human cancer types. Anti-SOX4 Antibody is ideal for research in Cell Differentiation and Development, Gene Expression, Apoptosis and Cancer.

Anti-SOX4 Antibody is suitable for ELISA and Western Blots. Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 47 kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: SOX4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR