Human Recombinant Sonic Hedgehog
Supplier: Rockland Immunochemical
009-001-W24-0100 009-001-W24-0025 009-001-W24-0005
75932-102EA 4452 USD
75932-102 75932-106 75932-110 75932-114
Human Recombinant Sonic Hedgehog
Proteins and Peptides
Recombinant Human Sonic Hedgehog control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Sonic hedgehog (SHH) is a member of a small group of secreted proteins that are essential for development in both vertebrates and invertebrates. Three mammalian hedgehog genes (sonic, desert, Indian) share about 60% homology and all signal via the same receptors. Recombinant human SHH is a non-glycosylated protein, containing 175 amino acids, with a molecular weight of 19.7 kDa. The Cys at position 25 has been substituted with Ile.
Sonic Hedgehog Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Sonic Hedgehog in immunological assays.