SOD1
Supplier: Rockland Immunochemical
009-001-T48S
75932-088EA 388.66 USD
75932-088 75932-090
SOD1
Proteins and Peptides
SOD1 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
SOD1 (superoxide dismutase 1) is the major soluble cytoplasmic enzyme responsible for destroying harmful free superoxide radicals in the body thereby providing defense against oxygen free-radical toxicity. Soluble cytoplasmic SOD1 is a copper- and zinc-containing enzyme and the SOD1 gene maps to chromosome 21q22 (1). Mutations in the SOD1 gene have been implicated to be the cause of familial amyotrophic lateral sclerosis, increased age-related muscle mass loss, early development of cataracts, macular degeneration, thymic involution, hepatocellular carcinoma, and shortened lifespan (2). SOD1 Protein is ideal for investigators involved in Signaling Proteins, Cell Stress & Chaperone Proteins, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Cellular Stress, Inflammation, and Neurobiology research.
SOD1 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. SOD1 Protein is suitable for use in Western Blot. Expect a band approximately ~45kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.