Anti-B303_sMgp1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-EH3
75931-810EA 635.22 USD
75931-810
Anti-B303_sMgp1 Rabbit Polyclonal Antibody
Antibodies
Severe fever with thrombocytopenia syndrome (SFTS) is an emerging infectious disease caused by SFTS virus, a newly discovered bunyavirus that can cause high rates of fatalities. This disease is thought to be transmitted through blood contact. The SFTS virus HB29 membrane glycoprotein polyprotein mRNA encodes two glycoproteins termed Phlebovirus glycoprotein G1 and G2 respectively. This antibody will detect the non-cleaved glycoprotein

Anti-SFTS Virus HB29 Antibody has been tested for use in ELISA. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 117 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: B303_sMgp1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR