Anti-NA Mouse Monoclonal Antibody [Clone: [10C5F12]]

Supplier: Rockland Immunochemical
200-301-EG1
75931-780EA 739.7 USD
75931-780
Anti-NA Mouse Monoclonal Antibody [Clone: [10C5F12]]
Antibodies
Influenza A virus has one of sixteen possible hemagglutinin (HA) surface proteins and one of nine possible neuraminidase (NA) surface proteins. In early 2009, a novel H1N1 swine-origin influenza (S-OIV) A virus was identified in specimens obtained from patients in Mexico and the United States. The genetic make-up of this swine flu virus is unlike any other: it is an H1N1 strain that combines a triple assortment first identified in 1998 including human, swine, and avian influenza with two new pig H3N2 virus genes from Eurasia, themselves of recent human origin.

Anti-Seasonal H1N1 Neuraminidase Antibody has been tested for use in ELISA. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 52 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NA
Clonality: Monoclonal
Clone: [10C5F12]
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR