SDS-PAGE Destain Solution, Rockland Immunochemicals
Supplier: Rockland Immunochemical
MB-022-1000
75931-768EA 97 USD
75931-768
SDS-PAGE Destain Solution, Rockland Immunochemicals
Electrophoresis Reagents Stain Removal Reagents
SDS-PAGE Destain Solution
  • ready-to-use as a working 1X solution
  • in-gel visualization of Coomassie Blue stained total protein
Destain Solution is a uniquely formulated, ready-to-use reagent specifically designed for in-gel visualization of Coomassie Blue stained total protein in polyacrylamide gels. The proprietary formulation of the solution ensures high detection sensitivity and low backgrounds.
This product is ready-to-use as a working 1X solution and requires no further dilution. This solution contains alcohol and acetic acid in a proprietary combination to enhance sensitivity/noise ratio.