Anti-RHOG Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-DY9
75931-596EA 635.22 USD
75931-596
Anti-RHOG Rabbit Polyclonal Antibody
Antibodies
The ras homolog family member G (RHOG), also known as rho-related GTP-binding protein, is a member of the Rho family of small GTPases, which cycle between inactive GDP-bound and active GTP-bound states and function as molecular switches in signal transduction cascades (1). RHOG controls a pathway that requires the microtubule network and activates Rac1 and Cdc42Hs independently of their growth factor signaling pathway (2). RHOG is also involved in the transcriptional regulation of interferon-gamma and nuclear factor of activated T cells (NFAT) and the regulation of the actin skeleton in lymphocytes (3).

Anti-RHOG Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 21 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: RHOG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR