Mouse Recombinant RELM beta (from E. coli)
Supplier: Rockland Immunochemical
010-001-W18-0100 010-001-W18-0025 010-001-W18-0005
75931-518EA 4582 USD
75931-518 75931-522 75931-526 75931-530
Mouse Recombinant RELM beta (from E. coli)
Proteins and Peptides
Recombinant Mouse RELM beta control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Resistin-Like Molecule-beta (RELM- β) is a member of a recently identified family of secreted proteins containing conserved cysteines in their C terminus. The RELM family consists of Resistin (also called FIZZ3), RELM-α (FIZZ1), and RELM-γ. Only Resistin and RELM-β have been identified in humans whereas all four RELM family members have been identified in rodents. Recombinant mouse RELM-β is a non-glycosylated protein, containing 83 amino acids, with a molecular weight of 8.9 kDa.
RELM beta Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-RELM beta in immunological assays.