Mouse Recombinant RELM alpha (from E. coli)
Supplier: Rockland Immunochemical
010-001-W17-0100 010-001-W17-0025 010-001-W17-0005
75931-508EA 4582 USD
75931-508 75931-510 75931-512 75931-514
Mouse Recombinant RELM alpha (from E. coli)
Proteins and Peptides
Recombinant Mouse RELM alpha control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Resistin-Like Molecule-alpha, or RELM-α, is a cytokine member to a unique family of proteins including Relm-β and Resistin. Relm-α and Resistin are secreted by adipocytes, where as Relm-β is secreted by intestinal epithelial cells. All members of the family share a high conserved C-terminal region with 10 Cysteine residues. Recombinant mouse RELM-α is a non-glycosylated protein, containing 88 amino acids, with a molecular weight of 10 kDa.
RELM alpha Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-RELM alpha in immunological assays.