Anti-PTGDR2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-DU6
75931-370EA 635.22 USD
75931-370
Anti-PTGDR2 Rabbit Polyclonal Antibody
Antibodies
The prostaglandin D2 receptor 2 (PTGDR2), also known as CRTH2, is expressed on Th2 cells and eosinophils and mediates chemotaxis of these cells to PGD2 and is thus thought to be a key receptor mediating eosinophil and Th2 recruitment during allergic responses. However, PTGDR2-null mice showed enhanced eosinophil recruitment into the lung consistent with observations that the PTGDR2-null mice produced significantly higher amounts of interleukin-5 (IL-5) and IL-3. This suggests that PTGDR2 plays a nonredundant role in restricting eosinophilia and allergic response in vivo.

Anti-PTGDR2 Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 43 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PTGDR2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR