Human recombinant procalcitonin (from E. coli)
Supplier: Rockland Immunochemical
009-001-W14-0010 009-001-W14-0100
75931-174EA 8502 USD
75931-174 75931-176 75931-178 75931-180
Human recombinant procalcitonin (from E. coli)
Proteins and Peptides
Recombinant Human Procalcitonin control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Procalcitonin is a precursor of the peptide hormone called calcitonin. Procalcitonin is mainly produced by the C cells of the thyroid and certain endocrine cells of the lung. Under normal expression conditions, procalcitonin is immediately cleaved into three specific fragments, an N terminal residue, calcitonin and katacalcin. Levels of unprocessed procalcitonin have been shown to rise significantly after bacterial infection, trauma or shock. Recombinant human Procalcitonin is a non-glycosylated protein, containing 116 amino acids, with a molecular weight of 12.8 kDa.
Procalcitonin Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Procalcitonin in immunological assays.