Anti-PLG Goat Polyclonal Antibody (BAC)

Supplier: Rockland Immunochemical
200-106-208S
75931-058EA 151.49 USD
75931-058
Anti-PLG Goat Polyclonal Antibody (BAC)
Antibodies
Anti-Plasminogen antibody recognizes plasminogen, a precursor of plasmin. Plasmin is a serine protease that degrades fibrin clots and involved in embryonic development, tissue remodeling, tumor invasion, and inflammation. In addition to fibrin, Plasmin can also cleave fibronectin, thrombospondin, laminin, and von Willebrand factor. Plasmin can activate other zymogens such as tissue plasminogen activator, urokinase plasminogen activator, kallikrein, and factor XII. Deficiency in plasminogen increases susceptibility to thrombosis.

Anti-Plasminogen antibody has been assayed against 1.0 ug of Plasminogen in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature.  A working dilution of 1:4,000 to 1:20,000 of the reconstitution concentration is suggested for this product.

Type: Primary
Antigen: PLG
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR