Anti-PIAS4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-DM9
75930-936EA 635.22 USD
75930-936
Anti-PIAS4 Rabbit Polyclonal Antibody
Antibodies
The PIAS (protein inhibitor of activated STAT) proteins play a crucial role as transcriptional coregulators in various cellular pathways, including the STAT, p53 and the steroid hormone signaling pathway. The PIAS protein family includes at least five evolutionarily conserved genes, including PIAS4. The major function of the PIAS proteins is the control of gene transcription and can also act as small ubiquitin-like-modifier (SUMO) E3 ligases. PIAS4 interacts with p53 and blocks its ability to induce Bax and p21 transcription. PIAS4 is also important in the control of the ubiquitin-dependent proteasomal degradation of the Ets-1 transcription factor. PIAS4 has been implicated in the DNA-damage response pathway and is thought to work in combination with PIAS1 for the productive association of 53BP1, BRCA1 and RNF168.

Anti-PIAS4 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 57 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PIAS4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR