PIK3R1
Supplier: Rockland Immunochemical
009-001-S93S
75930-914EA 380.3 USD
75930-914 75930-916
PIK3R1
Proteins and Peptides
PI3K (p65 alpha) control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PIK3R1 is a Phosphatidylinositol 3-kinase phosphorylates the inositol ring of phosphatidylinositol at the 3-prime position which comprises a 110 kD catalytic subunit and a regulatory subunit of either 85, 55, or 50 kD and it encodes the 85 kD regulatory subunit. PIK3R1 protein associates with activated growth factor receptors. p85-alpha modulates the interaction between PI3 kinase and platelet-derived growth factor receptor (1). PI3K plays an essential role in the development and induction of mast cells in normal and pathogenic immune responses (2).Phosphatidylinositol 3-kinase plays an important role in the metabolic actions of insulin, and a mutation in this gene has been associated with insulin resistance. PI3K (p65 alpha) Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, AKT/PKB Pathway, Angiogenesis, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Inflammation, Invasion/Metastasis, Lipid Kinases, Metabolic Disorder, Neurobiology, NfkB Pathway, and WNT Signaling research.
PI3K (p65 alpha) Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. PI3K (p65 alpha) Protein is suitable for use in Western Blot. Expect a band approximately ~67kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.