Anti-PHC3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-DM0
75930-864EA 635.22 USD
75930-864
Anti-PHC3 Rabbit Polyclonal Antibody
Antibodies
Polycomb group (PcG) proteins assemble into multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes throughout development (1). PHC3 (Polyhomeotic-like protein 3), also known as Early development regulatory protein 3, is a 983 amino acid nuclear protein that is a component of the PcG multiprotein PRC1 complex (2). PRC1 complex acts via chromatin remodeling and modification of histones. PHC3 contains 1 FCS-type zinc finger and SAM (sterile alpha motif) domain (2-3). PHC 3 has been regarded as a candidate tumor suppressor of osteosarcoma (4).

Anti-PHC3 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 106 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PHC3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR