Anti-FAM120B Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-DL4
75930-850EA 635.22 USD
75930-850
Anti-FAM120B Rabbit Polyclonal Antibody
Antibodies
The transcription factor peroxisome proliferator-activated receptor gamma (PPARg) plays essential roles in adipogenesis by regulating adipocyte-specific genes through association of various co-factors. One such co-factor, PGCC1 (also known as FAM120B), is widely expressed in adult tissues and throughout embryonic development. Overexpression of this protein in OP9 pre-adipocytes promoted their differentiation into adipocytes, and knockdown of PGCC1 expression through RNA interference blocked this process. PGCC1 is homologous to C9orf10 (also known as FAM120A) and has been mapped to chromosome Xp11.22. At least two isoforms of PGCC1 are known to exist.

Anti-PGCC1 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 104 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: FAM120B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR