CDK14
Supplier: Rockland Immunochemical
009-001-S90S
75930-846EA 388.66 USD
75930-846 75930-848
CDK14
Proteins and Peptides
PFTK1 (CDK14) control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PFTK1, also known as PFTAIRE protein kinase 1, is a member of the CDC2 -related protein kinase family which is expressed primarily in the postnatal and adult nervous system (1). PFTK1 is highly expressed in brain, pancreas, kidney, heart, testis, and ovary. PFTK1 interacts with 14-3-3-beta, 14-3-3-epsilon, 14-3-3-eta and 14-3-3-tau. Using PFTK1 mutant constructs and in vitro and in vivo binding studies, it was shown that PFTK1 amino acid residue ser119 is required for its interaction with all four 14-3-3 isoforms. (2). Significant upregulation of PFTK1 expression is observed in esophageal squamous cell carcinoma (ESCC). PFTK1 is not only useful as a prognostic marker in ESCC, but also as a predictor of the response to chemotherapy. PFTK1 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cancer, Cell Cycle, and Ser/Thr Kinases research.
PFTK1 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. PFTK1 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~72kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.