Anti-Perp Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-H10
75930-796EA 635.22 USD
75930-796
Anti-Perp Rabbit Polyclonal Antibody
Antibodies
PERP antibody detects human PERP. The p53 tumor-suppressor gene integrates numerous signals that control cell life and death. Several novel molecules involved in p53 network, including Chk2, p53R2, p53AIP1, Noxa, PIDD, PID/MTA2, MTBP and PERP, were identified and their genes were cloned recently. PERP, also termed PIGPC1 and THW, is a plasma membrane protein. p53 binds to the promoter of PERP and transcriptionally activates PERP gene then the translated PERP protein mediates the p53 induced apoptosis. The expression of PERP causes cell death. PERP is a mediator of p53 induced apoptosis. PERP has sequence similarity to PMP-22/gas3 and is a new member of the PMP-22/gas3 family. Anti-PERP antibodies are ideal for investigators involved in Apoptosis research.

Anti-PERP Antibody is suitable for use in E,WB, and ICC. Expect a band approximately ~21.5 kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: Perp
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR