Anti-PPHLN1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-DL3
75930-794EA 635.22 USD
75930-794
Anti-PPHLN1 Rabbit Polyclonal Antibody
Antibodies
Periphilin, known as PPHLN1 or gastric cancer antigen Ga50, plays a role in epithelial differentiation and contributes to epidermal integrity and barrier formation. It interacts with periplakin, a known precursor of the cornified cell envelope. Periphilin 1 is ubiquitously expressed and localizes to nucleus and cytoplasm. Existing as eight alternatively spliced isoforms, Periphilin is highly insoluble and contains a putative nuclear localization signal (NLS) within its N-terminal half, a prerequisite for the formation of insoluble complexes and a possible caspase recognition sequence and a potential nuclear export signal. Periphilin may play an important role in the nervous system.

Anti-Periphilin Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 53 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PPHLN1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR