CDK16
Supplier: Rockland Immunochemical
009-001-S83S
75930-754EA 388.66 USD
75930-754 75930-756
CDK16
Proteins and Peptides
PCTK1 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PCTK1 or Pctaire protein kinase 1 belongs to the CDC2/CDKX subfamily of the Ser/Thr family of protein kinases which play a main role in signal transduction cascades in terminally differentiated cells, in exocytosis and in transport of secretory cargo from the endoplasmic reticulum. PCTK1 is ubiquitously expressed with the highest levels detected in the brain and testis (1). PCTK1 maps distal to the t(X;18) synovial sarcoma breakpoint in Xp11.23. A 420-kb YAC clone positive for PCTK1 also contained the gene coding for ubiquitin-activating enzyme UBE1 previously mapped to Xp11.3. (2). PCTK1 is thought to escape X inactivation. PCTK1 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cancer, Cell Cycle, and Ser/Thr Kinases research.
PCTK1 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. PCTK1 Protein is suitable for use in Western Blot. Expect a band approximately ~62kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.