Anti-NBN Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-DB0
75930-428EA 635.22 USD
75930-428
Anti-NBN Rabbit Polyclonal Antibody
Antibodies
NIBRIN (NBN) is a member of the double-strand break repair complex MRE11/RAD50/NBN (MRN) which is involved in DNA double-strand break repair, DNA damage-induced checkpoint activation and plays a critical role in the maintenance of chromosome integrity. NIBRIN contains two modules found in cell cycle checkpoint proteins, a forkhead-associated domain adjacent to a breast cancer carboxy-terminal domain. Mutations in this gene are associated with Nijmegen breakage syndrome and maybe the cause of cancer predisposition and aplastic anemia.

Anti-NIBRIN Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 85 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NBN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR