Anti-NEUROG2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-DA6
75930-420EA 635.22 USD
75930-420
Anti-NEUROG2 Rabbit Polyclonal Antibody
Antibodies
Neurogenin-2 (NGN2) is a neural-specific basic helix-loop-helix (bHLH) transcription factor that can specify a neuronal fate on ectodermal cells and is expressed in neural progenitor cells within the developing central and peripheral nervous systems. NGN2 is thought to work with Nurr1 to play a role in the differentiation and survival of midbrain dopaminergic neurons. It has also been suggested for use in human embryonic neural progenitors as a graft for spinal cord injuries

Anti-NGN2 Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 29 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NEUROG2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR