Anti-NGFR Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-DA5
75930-418EA 635.22 USD
75930-418
Anti-NGFR Rabbit Polyclonal Antibody
Antibodies
The tumor necrosis factor (TNF) and TNF receptor (TNFR) gene superfamilies regulate numerous biological functions including cell proliferation, differentiation, and survival through regulating the activation of the transcription factor NF-kB and various mitogen-activated protein kinases. Nerve growth factor receptor (NGFR) was one of the earliest characterized members of this family. Also known as the low-affinity receptor p75NTR, this receptor is involved in several diverse functions such as apoptosis, neurite outgrowth during development, and myelination. Its ligands include NGF, brain-derived neurotrophic factor (BDNF), NT3, and NT4. NGFR can also associate with other NGF receptors such as Trk through the cytosolic and transmembrane domains and thus can function as a co-receptor that refines Trk affinity and specificity for neurotrophins. Finally, upon binding of various neurotrophins, NGFR associates with tumor necrosis factor receptor-6 (TRAF6), suggesting that it can potentially function as a signal transducer for NGF signals through NGFR.

Anti-NGFR Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 45 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NGFR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR