Anti-Neuroligin-1 Mouse Monoclonal Antibody (PerCP) [Clone: S97A-31]

Supplier: Rockland Immunochemical
200-365-W75
75930-338EA 589.25 USD
75930-338
Anti-Neuroligin-1 Mouse Monoclonal Antibody (PerCP) [Clone: S97A-31]
Antibodies
Neuroligin-1 is a neuronal cell surface protein belonging to the type-B carboxylesterase/lipase family. It is a necessary component in the maturation of excitatory synapses for their normal, functional development, and is necessary to the regulation of synaptic plasticity and the development of long-term memory within the adult amygdala in mammals. It is believed to participate in cell-cell-interaction through the assembly of intracellular junction by the binding of beta-neurexins, and may also be a factor in the maintenance and assembly of synaptic junctions. It is also thought to have involvement in excitatory synaptic specification. Within brain tissue, Neuroligin-1 is primarily observed in neurons and spinal cord. Anti-Neuroligin-1 is ideal for research in Neuroscience, including narcolepsy and synaptic neurotransmission: GABAergic inhibition, as well as Cell Adhesion.

Anti-Neuroligin 1 PerCP Conjugated Antibody is suitable for Western Blots, Immunohistochemistry and Immunocytochemistry. Expect a band approximately ~120kDa on specific lysates or tissues. Does not cross react with other Neuroligins. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: Neuroligin-1
Clonality: Monoclonal
Clone: S97A-31
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR