Anti-NEFM Mouse Monoclonal Antibody [Clone: 3H11]

Supplier: Rockland Immunochemical
912-301-D84
75930-308EA 578.81 USD
75930-308
Anti-NEFM Mouse Monoclonal Antibody [Clone: 3H11]
Antibodies
NF-M antibody detects Neurofilament M protein. Neurofilaments are the 10nm or intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. NF-M is the neurofilament middle or medium molecular weight polypeptide and runs on SDS-PAGE gels at 145-160 kDa, with some variability across species boundaries. Antibodies to NF-M are useful for identifying neuronal cells and their processes in tissue sections and in tissue culture. NF-M antibodies can also be useful to visualize neurofilament accumulations seen in many neurological diseases, such as Amyotrophic Lateral Sclerosis (Lou Gehrig's disease) and Alzheimer's disease. Anti-NF-M antibody is ideal for investigators involved in Cell Signaling and Neuroscience research.

Anti-NFM antibody is suitable for use in Western Blotting and IF. Expect a band of approximately 145 kDa in size corresponding to the NF-M proteins in the appropriate cell lysate or extract. Researchers should determine optimal titers for applications that are not stated below.

Type: Primary
Antigen: NEFM
Clonality: Monoclonal
Clone: 3H11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR