Anti-C1orf187 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-DA1
75930-294EA 635.22 USD
75930-294
Anti-C1orf187 Rabbit Polyclonal Antibody
Antibodies
Neucrin, also known as Draxin, is a repulsive guidance protein for the spinal cord and forebrain commissures. It is thought to act as an antagonist of canonical Wnt signaling by inhibiting the stabilization of cytosolic beta-catenin. Ectopically expressed neucrin inhibited growth or caused misrouting of chick spinal cord commissural axons in vivo while Neucrin-null mice showed defasciculation of spinal cord commissural axons and an absence of all forebrain commisures. Other experiments in mice have shown that olfactory bulb axonal outgrowth is inhibited by Neucrin, suggesting that Neucrin functions as an inhibitory guidance cue for olfactory bulb axons.

Anti-NEUCRIN Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 39 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: C1orf187
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR