Anti-STXBP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-X15
75930-162EA 656.12 USD
75930-162
Anti-STXBP1 Rabbit Polyclonal Antibody
Antibodies
Munc-18 (mammalian homologue of Unc-18) is a protein that is thought to be involved in regulating exocytosis due, at least in part, to it ability to bind syntaxin. Munc18-1 is a neuron-specific member of the Sec1/Munc18 protein family that binds to syntaxin1A and is thought to stabilize the complex. The function of Munc18-1 is thought to be regulated by phosphorylation. Munc18-1 pS241 Antibody is ideal for researchers interested in neuroscience research.

Anti-Phospho-Ser241 Munc18-1 Antibody is stored with 100 μg per ml BSA. It is suitable for Western Blots and is specific for the ~65 kDa Munc18-1 protein phosphorylated at Ser241. Immunolabeling is blocked by the treatment of the lysate with lambda phosphatase (30 minutes, 800 units / 1 mg protein). Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: STXBP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR